Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014037.2 | NP_054756.2 | 5 | 957 | ||
XM_005258820.2 | XP_005258877.1 | 6 | 1058 | |||
XM_006723168.3 | XP_006723231.1 | 5 | 973 | |||
XM_011526859.2 | XP_011525161.2 | 5 | 991 | |||
XM_011526860.2 | XP_011525162.1 | 6 | 1270 | |||
XM_011526861.2 | XP_011525163.1 | 4 | 380 | |||
XM_017026712.1 | XP_016882201.1 | 5 | 973 | |||
GenBank mRNA | AF151977.1 | - | 4 | 762 | ||
AF265578.1 | - | 4 | 723 | |||
AL136856.1 | - | 4 | 769 | |||
BC034948.1 | - | 5 | 925 | 4821848 |