Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052934.3 | NP_443166.1 | 5 | 586 | ||
NM_134325.2 | NP_599152.2 | 5 | 586 | |||
XM_011509121.2 | XP_011507423.1 | 4 | 449 | |||
XM_011509122.2 | XP_011507424.1 | 2 | 138 | |||
XM_011509124.2 | XP_011507426.1 | 5 | 598 | |||
GenBank mRNA | AF314958.1 | - | 4 | 474 | ||
AF331525.1 | - | 5 | 586 | |||
BC136538.1 | - | 5 | 566 | 9020528 | ||
BC144233.1 | - | 4 | 455 | |||
BC151208.1 | - | 5 | 545 | 40147148 |