Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC23A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005847.4 | NP_005838.3 | 10 | 1190 | ||
NM_152685.3 | NP_689898.2 | 10 | 1202 | |||
XM_005272148.3 | XP_005272205.3 | 11 | 1327 | |||
XM_005272149.3 | XP_005272206.3 | 11 | 1315 | |||
XM_006714741.2 | XP_006714804.2 | 11 | 1327 | |||
XM_011543765.2 | XP_011542067.1 | 11 | 1327 | |||
XM_011543766.1 | XP_011542068.1 | 10 | 1108 | |||
XM_011543767.1 | XP_011542069.1 | 9 | 1012 | |||
XM_011543768.1 | XP_011542070.1 | 10 | 1232 | |||
XM_011543769.1 | XP_011542071.1 | 9 | 987 | |||
GenBank mRNA | AF058317.1 | - | 10 | 1093 | ||
AF098277.1 | - | 10 | 1140 | |||
AF170911.1 | - | 10 | 1124 | |||
AJ250807.1 | - | 10 | 1163 | |||
AJ269477.1 | - | 10 | 1093 | |||
AK092655.1 | - | 9 | 1014 | |||
AK290789.1 | - | 10 | 1123 | |||
AK298337.1 | - | 9 | 1023 | |||
BC050261.1 | - | 10 | 1144 | 5188430 |