Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145975.1 | NP_001139447.1 | 3 | 534 | ||
NM_003984.3 | NP_003975.1 | 3 | 387 | |||
NR_027384.1 | 4 | 528 | ||||
XM_006722165.2 | XP_006722228.1 | 4 | 436 | |||
XM_011525450.2 | XP_011523752.1 | 3 | 800 | |||
XM_011525451.1 | XP_011523753.1 | 4 | 596 | |||
XM_011525452.1 | XP_011523754.1 | 3 | 387 | |||
XM_011525453.2 | XP_011523755.1 | 3 | 808 | |||
XM_011525454.2 | XP_011523756.1 | 3 | 822 | |||
GenBank mRNA | AK091926.1 | - | 5 | 498 | ||
AK298388.1 | - | 3 | 442 | |||
AK303909.1 | - | 4 | 436 | |||
AK303936.1 | - | 2 | 166 | |||
AK314684.1 | - | 3 | 387 | |||
BC096276.1 | - | 4 | 482 | 40006564 | ||
BC096277.1 | - | 3 | 341 | 40006565 | ||
BC096278.1 | - | 3 | 341 | 40006566 | ||
BC096279.1 | - | 3 | 341 | 40006572 | ||
DB204108.1 | - | 2 | 166 | |||
U26209.1 | - | 3 | 388 |