Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136019.2 | NP_001129491.1 | 3 | 782 | ||
NM_004107.4 | NP_004098.1 | 2 | 518 | |||
XM_005258657.1 | XP_005258714.1 | 2 | 337 | |||
XM_011526627.2 | XP_011524929.1 | 2 | 475 | |||
GenBank mRNA | AK074734.1 | - | 1 | 909 | ||
AK075510.1 | - | 2 | 329 | |||
AK075532.1 | - | 3 | 432 | |||
AK299124.1 | - | 2 | 302 | |||
BC008734.2 | - | 2 | 462 | 3163446 | ||
BC020421.1 | - | 1 | 715 | |||
BT006828.1 | - | 2 | 296 | |||
BX647163.1 | - | 3 | 402 | |||
DC312637.1 | - | 2 | 519 | |||
U12255.1 | - | 2 | 421 |