Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184732.1 | NP_001171661.1 | 2 | 437 | ||
NM_001184733.1 | NP_001171662.1 | 2 | 185 | |||
NM_004254.3 | NP_004245.2 | 2 | 458 | |||
GenBank mRNA | AB042505.1 | - | 2 | 440 | ||
AF097491.1 | - | 1 | 316 | |||
AK298341.1 | - | 2 | 155 | |||
AK298351.1 | - | 3 | 410 | |||
AK313187.1 | - | 2 | 431 | |||
BC022387.1 | - | 2 | 452 | 4608542 | ||
BP276255.1 | - | 2 | 155 | |||
DA628705.1 | - | 2 | 458 | |||
DA633836.1 | - | 2 | 407 |