Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004790.4 | NP_004781.2 | 5 | 1232 | ||
NM_153276.2 | NP_695008.1 | 5 | 1232 | |||
NM_153277.2 | NP_695009.1 | 5 | 1232 | |||
NM_153278.2 | NP_695010.1 | 5 | 1232 | |||
XM_017018562.1 | XP_016874051.1 | 5 | 1180 | |||
GenBank mRNA | AB009697.1 | - | 5 | 1166 | ||
AB009698.1 | - | 5 | 1163 | |||
AF057039.2 | - | 5 | 1143 | |||
AF097490.1 | - | 5 | 899 | |||
AF104038.1 | - | 5 | 1140 | |||
AF124373.1 | - | 5 | 1156 | |||
AJ251529.1 | - | 5 | 899 | |||
AJ271205.1 | - | 5 | 899 | |||
AK055764.1 | - | 6 | 1145 | |||
AK091879.1 | - | 5 | 1157 | |||
AK298374.1 | - | 6 | 1042 | |||
BC033682.1 | - | 5 | 1192 | 5190748 |