Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256534.1 | NP_001243463.1 | 3, 4 | 613 | ||
NM_001287387.1 | NP_001274316.1 | 4, 5 | 467 | |||
NM_005984.4 | NP_005975.1 | 4, 5 | 632 | |||
GenBank mRNA | AK292313.1 | - | 4, 5 | 536 | ||
AK298204.1 | - | 4, 5 | 465 | |||
BC004980.1 | - | 4, 5 | 493 | 2957917 | ||
BC008061.2 | - | 4, 5 | 495 | 2958945 | ||
BC018590.1 | - | 4, 5 | 390 | |||
BG473148.1 | - | 3, 4 | 587 | |||
DR006840.1 | - | 4, 5 | 534 | |||
HM037273.1 | - | 3, 4 | 468 | |||
L75823.1 | - | 4, 5 | 541 | |||
L77567.1 | - | 4, 5 | 514 | |||
U25147.1 | - | 4, 5 | 516 |