Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190997.2 | NP_001177926.1 | 5 | 597 | ||
NM_016615.4 | NP_057699.2 | 7 | 873 | |||
XM_006719008.3 | XP_006719071.1 | 2 | 660 | |||
XM_011521012.2 | XP_011519314.1 | 7 | 941 | |||
XM_011521013.1 | XP_011519315.1 | 8 | 1009 | |||
XM_011521014.1 | XP_011519316.1 | 6 | 765 | |||
XM_017019842.1 | XP_016875331.1 | 2 | 666 | |||
XM_017019843.1 | XP_016875332.1 | 7 | 836 | |||
XM_017019844.1 | XP_016875333.1 | 7 | 836 | |||
XM_017019845.1 | XP_016875334.1 | 3 | 258 | |||
XM_017019846.1 | XP_016875335.1 | 7 | 836 | |||
XM_017019847.1 | XP_016875336.1 | 7 | 836 | |||
GenBank mRNA | AK295904.1 | - | 7 | 698 | ||
AK296127.1 | - | 5 | 560 | |||
AK296209.1 | - | 8 | 804 | |||
AK313511.1 | - | 7 | 829 | |||
BC022392.1 | - | 7 | 836 | 4612245 | ||
U76343.2 | - | 7 | 748 |