Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017767.2 | NP_060237.2 | 4 | 1083 | ||
NM_130849.3 | NP_570901.2 | 5 | 939 | |||
XM_006716599.1 | XP_006716662.1 | 5 | 918 | |||
GenBank mRNA | AK000334.1 | - | 4 | 1063 | ||
AK000695.1 | - | 4 | 998 | |||
AK025537.1 | - | 5 | 933 | |||
AK290563.1 | - | 5 | 887 | |||
BC062625.1 | - | 5 | 919 | 5203858 | ||
KU178707.1 | - | 5 | 833 | |||
KU178708.1 | - | 4 | 551 |