Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPSAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243940.1 | NP_001230869.1 | 3, 4 | 295 | ||
NM_002767.3 | NP_002758.1 | 4, 5 | 489 | |||
XM_005256725.2 | XP_005256782.1 | 3, 4 | 343 | |||
XM_005256726.1 | XP_005256783.1 | 3, 4 | 377 | |||
XM_017024864.1 | XP_016880353.1 | 4, 5 | 353 | |||
XM_017024865.1 | XP_016880354.1 | 5, 6 | 562 | |||
XM_017024866.1 | XP_016880355.1 | 5, 6 | 496 | |||
XM_017024867.1 | XP_016880356.1 | 3, 4 | 330 | |||
XM_017024868.1 | XP_016880357.1 | 4, 5 | 1207 | |||
XM_017024869.1 | XP_016880358.1 | 4, 5 | 486 | |||
XM_017024870.1 | XP_016880359.1 | 4, 5 | 401 | |||
GenBank mRNA | AB007851.1 | - | 3, 4 | 254 | ||
AL535304.3 | - | 4, 5 | 386 | |||
BC101670.1 | - | 3, 4 | 215 | 8069176 | ||
BC101672.1 | - | 3, 4 | 215 | 8069178 | ||
BC143475.1 | - | 3, 4 | 215 | 9051983 | ||
DA257655.1 | - | 3, 4 | 377 | |||
DC424578.1 | - | 4, 5 | 489 |