Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006045.2 | NP_006036.1 | 28 | 3248 | ||
XM_011528481.2 | XP_011526783.1 | 28 | 3276 | |||
GenBank mRNA | AB014511.1 | - | 23 | 2802 | ||
AK172802.1 | - | 26 | 2938 | |||
AK299064.1 | - | 28 | 3177 | |||
BC110591.1 | - | 24 | 2740 | |||
BC110592.1 | - | 24 | 2740 |