Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000702.3 | NP_000693.1 | 4, 5 | 507 | ||
GenBank mRNA | AB018321.2 | - | 4, 5 | 464 | ||
AK091617.1 | - | 4, 5 | 478 | |||
AK314296.1 | - | 4, 5 | 478 | |||
AL831991.1 | - | 3, 4 | 296 | |||
AL831997.1 | - | 3, 4 | 322 | |||
AY946014.1 | - | 1, 2 | 130 | |||
BC052271.1 | - | 4, 5 | 458 | 6672133 | ||
DA221602.1 | - | 4, 5 | 507 | |||
KU177927.1 | - | 3, 4 | 375 | |||
KU177928.1 | - | 3, 4 | 375 |