Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000702.3 | NP_000693.1 | 23 | 3561 | ||
GenBank mRNA | AB018321.2 | - | 23 | 3518 | ||
AK124581.1 | - | 21 | 3233 | |||
AK126573.1 | - | 16 | 2510 | |||
AL831991.1 | - | 22 | 3350 | |||
AL831997.1 | - | 22 | 3376 | |||
BC052271.1 | - | 23 | 3512 | 6672133 |