Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000702.3 | NP_000693.1 | 23 | 5395 | ||
GenBank mRNA | AB018321.2 | - | 23 | 5351 | ||
AL831991.1 | - | 22 | 5182 | |||
AL831997.1 | - | 22 | 5208 | |||
BC013680.1 | - | 1 | 712 |