Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098.2 | NP_001089.1 | 2 | 144 | ||
XM_017028812.1 | XP_016884301.1 | 2 | 458 | |||
GenBank mRNA | AK296260.1 | - | 2 | 147 | ||
AK297213.1 | - | 2 | 152 | |||
AK301321.1 | - | 2, 3 | 152 | |||
AK302691.1 | - | 2 | 151 | |||
AK314845.1 | - | 2 | 152 | |||
BC014092.2 | - | 2 | 144 | 4328775 | ||
BC026196.2 | - | 2 | 147 | 5264179 | ||
CR456365.1 | - | 2 | 136 | |||
CR536568.1 | - | 2 | 122 | |||
GQ472196.1 | - | 2 | 147 | |||
U80040.1 | - | 2 | 142 |