Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253846.1 | NP_001240775.1 | 6, 7 | 612 | ||
NM_001500.3 | NP_001491.1 | 6, 7 | 866 | |||
XM_006715066.2 | XP_006715129.1 | 6, 7 | 822 | |||
XM_011514500.1 | XP_011512802.1 | 6, 7 | 579 | |||
XM_011514502.2 | XP_011512804.1 | 6, 7 | 822 | |||
XM_017010752.1 | XP_016866241.1 | 3, 4 | 1095 | |||
GenBank mRNA | AF040260.1 | - | 6, 7 | 612 | ||
AF042377.1 | - | 6, 7 | 729 | |||
AK057484.1 | - | 1 | 780 | |||
AK313953.1 | - | 6, 7 | 806 | |||
AL561275.3 | - | 2, 3 | 314 | |||
BC000117.1 | - | 6, 7 | 831 | 3351070 | ||
CR541929.1 | - | 6, 7 | 654 | |||
CR541947.1 | - | 6, 7 | 654 |