Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313941.1 | NP_001300870.1 | 6 | 930 | ||
NM_001313943.1 | NP_001300872.1 | 6 | 974 | |||
NM_001313944.1 | NP_001300873.1 | 5 | 775 | |||
NM_001313945.1 | NP_001300874.1 | 5 | 712 | |||
NM_001313946.1 | NP_001300875.1 | 3 | 583 | |||
NM_001313947.1 | NP_001300876.1 | 4 | 714 | |||
NM_001664.3 | NP_001655.1 | 5 | 835 | |||
GenBank mRNA | AF498970.1 | - | 4 | 555 | ||
AK130066.1 | - | 5 | 831 | |||
AK130808.1 | - | 5 | 711 | |||
AK222556.1 | - | 5 | 727 | |||
AK296654.1 | - | 5 | 651 | |||
AK307220.1 | - | 5 | 610 | |||
BC000946.3 | - | 2 | 279 | |||
BC001360.2 | - | 5 | 667 | 3051460 | ||
BC005976.1 | - | 5 | 672 | 4102976 | ||
BT019870.1 | - | 4 | 555 | |||
BU509012.1 | - | 4 | 545 | |||
BX647063.1 | - | 5 | 731 | |||
L09159.1 | - | 4 | 544 | |||
L25080.1 | - | 5 | 706 | |||
LN607834.1 | - | 2 | 281 | |||
X05026.1 | - | 5 | 713 |