Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131061.1 | - | 3 | 426 | ||
AK293697.1 | - | 4 | 521 | |||
BC011835.2 | - | 3 | 418 | 3944210 | ||
BI462090.1 | - | 3 | 458 | |||
U51478.1 | - | 3 | 295 |