Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002804.4 | NP_002795.2 | 9, 10 | 1173 | ||
XM_017018026.1 | XP_016873515.1 | 9, 10 | 1173 | |||
GenBank mRNA | AK222485.1 | - | 9, 10 | 935 | ||
AK291896.1 | - | 9, 10 | 950 | |||
AK313518.1 | - | 9, 10 | 1159 | |||
BC008713.2 | - | 9, 10 | 1137 | 2822002 | ||
BC073165.1 | - | 9, 10 | 1118 | 6195828 | ||
BC106920.2 | - | 9, 10 | 1065 | 40032717 | ||
BC107804.1 | - | 9, 10 | 1137 | |||
CR456731.1 | - | 8, 9 | 874 | |||
M34079.1 | - | 9, 10 | 936 |