Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199163.1 | NP_001186092.1 | 5 | 518 | ||
NM_002805.5 | NP_002796.4 | 5 | 383 | |||
GenBank mRNA | AF035309.1 | - | 461 | |||
AK095836.1 | - | 4 | 1617 | |||
AK098484.1 | - | 1466 | ||||
AK290758.1 | - | 5 | 481 | |||
AK291878.1 | - | 5 | 355 | |||
BC001932.1 | - | 5 | 334 | 2820259 | ||
BC002367.2 | - | 5 | 313 | 2821416 | ||
BC127888.1 | - | 3 | 225 | |||
BF245599.1 | - | 368 | ||||
D44467.1 | - | 5 | 313 | |||
L38810.1 | - | 5 | 320 |