Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199163.1 | NP_001186092.1 | 8 | 939 | ||
NM_002805.5 | NP_002796.4 | 8 | 804 | |||
GenBank mRNA | AF035309.1 | - | 1225 | |||
AK095836.1 | - | 7 | 2038 | |||
AK098484.1 | - | 2068 | ||||
AK290758.1 | - | 8 | 902 | |||
AK291878.1 | - | 8 | 776 | |||
BC001932.1 | - | 8 | 755 | 2820259 | ||
BC002367.2 | - | 8 | 734 | 2821416 | ||
BC127888.1 | - | 6 | 646 | |||
D44467.1 | - | 8 | 734 | |||
L38810.1 | - | 8 | 741 |