Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002865.2 | NP_002856.1 | 2, 3 | 414 | ||
GenBank mRNA | AF498930.1 | - | 2, 3 | 116 | ||
AK312344.1 | - | 2, 3 | 337 | |||
BC008929.2 | - | 2, 3 | 304 | 2966694 | ||
BT019695.1 | - | 2, 3 | 116 | |||
DA648682.1 | - | 2, 3 | 416 | |||
M28213.1 | - | 2, 3 | 156 | |||
X12953.1 | - | 2, 3 | 324 | |||
X13899.1 | - | 2, 3 | 116 |