Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002890.2 | NP_002881.1 | 2 | 756 | ||
NM_022650.2 | NP_072179.1 | 2 | 150 | |||
XM_011543525.1 | XP_011541827.1 | 2 | 1208 | |||
XM_011543527.2 | XP_011541829.1 | 2 | 1208 | |||
GenBank mRNA | AB209106.1 | - | 2 | 756 | ||
AK300263.1 | - | 3 | 257 | |||
AK300269.1 | - | 2 | 83 | |||
AK300406.1 | - | 3 | 176 | |||
AK312739.1 | - | 2 | 135 | |||
BC020761.1 | - | 2 | 137 | |||
BC033015.2 | - | 2 | 744 | 4829173 | ||
BC054891.1 | - | 2 | 116 | |||
BC107870.1 | - | 2 | 140 | |||
CR749722.1 | - | 2 | 1121 | |||
M23379.1 | - | 2 | 675 | |||
M23612.1 | - | 2 | 125 |