Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202498.1 | NP_001189427.1 | 7 | 508 | ||
NM_003344.3 | NP_003335.1 | 7 | 904 | |||
NM_182697.2 | NP_874356.1 | 5 | 811 | |||
GenBank mRNA | AB451490.1 | - | 7 | 499 | ||
AK091088.1 | - | 1 | 852 | |||
AK316590.1 | - | 7 | 487 | |||
AL832212.1 | - | 1 | 913 | |||
BC006277.2 | - | 7 | 816 | 3957107 | ||
BI088146.1 | - | 7 | 508 | |||
BT006756.1 | - | 7 | 499 | |||
KJ534959.1 | - | 7 | 499 | |||
Z29328.1 | - | 7 | 552 | |||
Z29330.1 | - | 7 | 552 | |||
Z29331.1 | - | 2 | 169 |