Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003345.4 | NP_003336.1 | 5 | 401 | ||
NM_194259.2 | NP_919235.1 | 6 | 658 | |||
NM_194260.2 | NP_919236.1 | 5 | 521 | |||
NM_194261.2 | NP_919237.1 | 5 | 373 | |||
XM_005255540.2 | XP_005255597.1 | 5 | 392 | |||
XM_005255541.4 | XP_005255598.1 | 5 | 798 | |||
XM_011522644.2 | XP_011520946.1 | 6 | 638 | |||
XM_011522645.1 | XP_011520947.1 | 5 | 503 | |||
XM_017023640.1 | XP_016879129.1 | 5 | 798 | |||
GenBank mRNA | AB208988.1 | - | 5 | 482 | ||
AJ002385.1 | - | 5 | 321 | |||
AK024172.1 | - | 1 | 939 | |||
AK291118.1 | - | 5 | 392 | |||
BC000427.2 | - | 5 | 305 | 2819764 | ||
BC004429.1 | - | 5 | 305 | 2819764 | ||
BC051289.1 | - | 5 | 335 | |||
BE262845.1 | - | 6 | 640 | |||
BI523501.1 | - | 5 | 511 | |||
BT006932.1 | - | 4 | 243 | |||
BT007010.1 | - | 4 | 243 | |||
BU543455.1 | - | 5 | 393 | |||
CD249028.1 | - | 5 | 342 | |||
CX788607.1 | - | 5 | 477 | |||
D45050.1 | - | 5 | 360 | |||
U29092.1 | - | 4 | 267 | |||
U31882.1 | - | 5 | 336 | |||
U31933.1 | - | 5 | 361 | |||
U38785.1 | - | 5 | 335 | |||
U45328.1 | - | 5 | 366 | |||
U66818.1 | - | 5 | 330 | |||
U66867.1 | - | 5 | 1049 | |||
X96427.1 | - | 5 | 366 |