Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003350.2 | NP_003341.1 | 4 | 923 | ||
XM_005251300.2 | XP_005251357.1 | 4 | 982 | |||
XM_011517583.2 | XP_011515885.1 | 4 | 1001 | |||
XM_017013808.1 | XP_016869297.1 | 4 | 908 | |||
GenBank mRNA | AF049140.1 | - | 4 | 923 | ||
AK094617.1 | - | 3 | 2172 | |||
BC016332.1 | - | 4 | 948 | 4109228 | ||
BC016710.1 | - | 4 | 932 | 4108684 | ||
BC028673.1 | - | 4 | 925 | 4104992 | ||
BC062418.1 | - | 4 | 922 | 6503261 | ||
U62136.2 | - | 4 | 924 | |||
X98091.1 | - | 4 | 948 |