Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003356.3 | NP_003347.1 | 2 | 447 | ||
NM_022803.2 | NP_073714.1 | 2 | 447 | |||
GenBank mRNA | AF001787.1 | - | 2 | 275 | ||
AF011449.1 | - | 1 | 190 | |||
AK290630.1 | - | 2 | 295 | |||
AK291889.1 | - | 2 | 261 | |||
AK309873.1 | - | 2 | 295 | |||
BC008392.1 | - | 2 | 295 | 4291158 | ||
DA876282.1 | - | 2 | 447 | |||
KU178371.1 | - | 1 | 92 | |||
KU178372.1 | - | 1 | 92 | |||
U82818.1 | - | 2 | 245 | |||
U84763.1 | - | 2 | 245 |