Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135662.1 | NP_001129134.1 | 4 | 470 | ||
NM_001135663.1 | NP_001129135.1 | 2 | 685 | |||
NM_001135664.1 | NP_001129136.1 | 3 | 317 | |||
NM_003929.2 | NP_003920.1 | 4 | 571 | |||
XM_005245569.1 | XP_005245626.1 | 4 | 469 | |||
XM_005245570.1 | XP_005245627.1 | 4 | 522 | |||
XM_005245571.1 | XP_005245628.1 | 4 | 447 | |||
XM_006711605.3 | XP_006711668.1 | 3 | 213 | |||
XM_006711606.2 | XP_006711669.1 | 3 | 177 | |||
XM_017002748.1 | XP_016858237.1 | 3 | 472 | |||
XM_017002749.1 | XP_016858238.1 | 3 | 450 | |||
XM_017002750.1 | XP_016858239.1 | 3 | 382 | |||
GenBank mRNA | AK223514.1 | - | 3 | 347 | ||
AK303879.1 | - | 2 | 436 | |||
AK308359.1 | - | 3 | 286 | |||
AK313000.1 | - | 4 | 459 | |||
BC002585.2 | - | 4 | 404 | 3140744 | ||
BT019540.1 | - | 3 | 231 | |||
BX647960.1 | - | 4 | 536 | |||
CR542010.1 | - | 3 | 231 | |||
D84488.1 | - | 3 | 271 | |||
DA284582.1 | - | 4 | 470 | |||
DA377295.1 | - | 3 | 272 |