Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004040.3 | NP_004031.1 | 1 | 1985 | ||
GenBank mRNA | AF171089.1 | - | 1 | 471 | ||
AK124398.1 | - | 1 | 1982 | |||
BC066954.1 | - | 1 | 1982 | 5264057 |