Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004161.4 | NP_004152.1 | 3 | 562 | ||
NM_015543.1 | NP_056358.1 | 3 | 562 | |||
XM_005264468.1 | XP_005264525.1 | 3 | 367 | |||
GenBank mRNA | AF498929.1 | - | 3 | 176 | ||
AK303678.1 | - | 3 | 366 | |||
AK315495.1 | - | 3 | 371 | |||
AL050268.1 | - | 3 | 366 | |||
BC000905.2 | - | 3 | 354 | 2900705 | ||
BG704216.1 | - | 3 | 676 | |||
BT019528.1 | - | 3 | 176 | |||
BT019529.1 | - | 3 | 176 | |||
BX571747.1 | - | 3 | 359 | |||
CR533479.1 | - | 3 | 176 | |||
CR536488.1 | - | 3 | 176 | |||
M28209.1 | - | 3 | 226 | |||
X13898.1 | - | 3 | 176 |