Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004223.4 | NP_004214.1 | 3 | 252 | ||
NM_198183.2 | NP_937826.1 | 3 | 611 | |||
GenBank mRNA | AF031141.1 | - | 3 | 153 | ||
AF061736.1 | - | 3 | 203 | |||
AK093462.1 | - | 3 | 565 | |||
AK129621.1 | - | 3 | 123 | |||
AK226165.1 | - | 3 | 306 | |||
AK315755.1 | - | 3 | 171 | |||
BC032491.1 | - | 3 | 252 | 5246109 | ||
CR457126.1 | - | 3 | 156 |