Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004580.4 | NP_004571.2 | 6 | 849 | ||
NM_183234.2 | NP_899057.1 | 7 | 830 | |||
NM_183235.2 | NP_899058.1 | 7 | 839 | |||
NM_183236.2 | NP_899059.1 | 7 | 790 | |||
XM_005254576.4 | XP_005254633.1 | 6 | 2092 | |||
XM_011521852.1 | XP_011520154.1 | 9 | 1053 | |||
XM_011521854.1 | XP_011520156.1 | 8 | 926 | |||
XM_011521855.2 | XP_011520157.1 | 8 | 2315 | |||
XM_011521856.2 | XP_011520158.1 | 6 | 722 | |||
GenBank mRNA | AF125393.1 | - | 7 | 806 | ||
AF498953.1 | - | 5 | 597 | |||
AK312836.1 | - | 7 | 837 | |||
BC107680.1 | - | 7 | 790 | 5416688 | ||
BC132800.1 | - | 7 | 775 | 40146822 | ||
BC136423.1 | - | 7 | 775 | 9020412 | ||
CR457076.1 | - | 5 | 597 | |||
CR536496.1 | - | 5 | 597 | |||
CR541693.1 | - | 5 | 597 | |||
U38654.3 | - | 6 | 842 | |||
U57094.1 | - | 7 | 830 |