Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252039.1 | NP_001238968.1 | 7 | 1182 | ||
NM_004583.3 | NP_004574.2 | 6 | 966 | |||
NM_201434.2 | NP_958842.1 | 7 | 1083 | |||
XM_011525090.1 | XP_011523392.1 | 6 | 905 | |||
XM_011525091.1 | XP_011523393.1 | 6 | 1029 | |||
GenBank mRNA | AK310234.1 | - | 7 | 1126 | ||
BC106039.1 | - | 6 | 925 | 4938169 | ||
BC114439.1 | - | 6 | 921 | 40083572 | ||
U11293.1 | - | 5 | 801 | |||
U18420.1 | - | 6 | 861 |