Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004661.3 | NP_004652.2 | 5 | 525 | ||
GenBank mRNA | AB011472.1 | - | 5 | 512 | ||
AF053977.1 | - | 5 | 492 | |||
AF191341.1 | - | 5 | 476 | |||
AK223250.1 | - | 5 | 513 | |||
AK291610.1 | - | 5 | 507 | |||
AK304635.1 | - | 4 | 389 | |||
BC017713.1 | - | 5 | 484 | 4399056 | ||
BT009810.1 | - | 5 | 476 |