Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004661.3 | NP_004652.2 | 5 | 529 | ||
GenBank mRNA | AB011472.1 | - | 5 | 516 | ||
AF053977.1 | - | 5 | 496 | |||
AF191341.1 | - | 5 | 480 | |||
AK223250.1 | - | 5 | 517 | |||
AK291610.1 | - | 5 | 511 | |||
AK304635.1 | - | 4 | 393 | |||
BC017713.1 | - | 5 | 488 | 4399056 | ||
BT009810.1 | - | 5 | 480 |