Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243738.1 | NP_001230667.1 | 6 | 1236 | ||
NM_004761.4 | NP_004752.1 | 7 | 1433 | |||
XM_011514781.1 | XP_011513083.1 | 5 | 705 | |||
XM_011514785.2 | XP_011513087.1 | 5 | 705 | |||
GenBank mRNA | AB012295.1 | - | 6 | 785 | ||
AK095967.1 | - | 4 | 407 | |||
AK294442.1 | - | 6 | 772 | |||
AL050259.1 | - | 7 | 1006 | |||
BC000316.1 | - | 1 | 92 | |||
BC005037.1 | - | 4 | 474 | |||
BC032681.1 | - | 7 | 988 | 5585507 | ||
BC040989.1 | - | 6 | 1288 | |||
BX648015.1 | - | 6 | 778 | |||
U68142.1 | - | 1 | 39 |