Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254738.1 | NP_001241667.1 | 2 | 468 | ||
NM_005168.4 | NP_005159.1 | 3 | 375 | |||
GenBank mRNA | AF498969.1 | - | 2 | 190 | ||
AK298007.1 | - | 3 | 345 | |||
AK299731.1 | - | 2 | 175 | |||
AK313224.1 | - | 2 | 442 | |||
AL702945.1 | - | 2 | 441 | |||
BC012513.1 | - | 3 | 330 | 4453777 | ||
BQ440721.1 | - | 2 | 444 | |||
BT006769.1 | - | 2 | 190 | |||
CR542038.1 | - | 2 | 145 | |||
CR542070.1 | - | 2 | 145 | |||
DC367501.1 | - | 3 | 376 | |||
S82240.1 | - | 2 | 169 | |||
X95282.1 | - | 2 | 190 | |||
X97758.1 | - | 2 | 412 |