Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005402.3 | NP_005393.2 | 5 | 2649 | ||
XM_006715762.2 | XP_006715825.1 | 5 | 2441 | |||
XM_011515466.1 | XP_011513768.1 | 6 | 2693 | |||
GenBank mRNA | AK026850.1 | - | 1 | 1784 | ||
BC039858.1 | - | 5 | 2595 | 5495399 | ||
BM930625.1 | - | 1 | 239 |