Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098670.1 | NP_001092140.1 | 3, 4 | 360 | ||
NM_001098671.1 | NP_001092141.1 | 3, 4 | 400 | |||
NM_001318398.1 | NP_001305327.1 | 3, 4 | 320 | |||
NM_153819.1 | NP_722541.1 | 3, 4 | 426 | |||
XM_005273707.3 | XP_005273764.3 | 3, 4 | 751 | |||
XM_011544718.1 | XP_011543020.1 | 3, 4 | 751 | |||
XM_011544720.1 | XP_011543022.1 | 3, 4 | 831 | |||
XM_011544721.1 | XP_011543023.1 | 3, 4 | 363 | |||
XM_011544722.1 | XP_011543024.1 | 3, 4 | 416 | |||
XM_011544723.2 | XP_011543025.1 | 3, 4 | 365 | |||
XM_011544725.2 | XP_011543027.1 | 1, 2 | 115 | |||
XM_017017082.1 | XP_016872571.1 | 2, 3 | 1401 | |||
XM_017017083.1 | XP_016872572.1 | 2, 3 | 1401 | |||
XM_017017084.1 | XP_016872573.1 | 3, 4 | 394 | |||
GenBank mRNA | AF043722.1 | - | 4, 5 | 532 | ||
AF043723.1 | - | 3, 4 | 426 | |||
AF081194.1 | - | 3, 4 | 333 | |||
AK092882.1 | - | 2, 3 | 1096 | |||
AK122694.1 | - | 3, 4 | 284 | |||
AK124023.1 | - | 2, 3 | 1896 | |||
AK314637.1 | - | 2, 3 | 311 | |||
BC110306.1 | - | 3, 4 | 334 | |||
BC117151.1 | - | 3, 4 | 424 | 40125702 | ||
DA286186.1 | - | 3, 4 | 420 | |||
DA529151.1 | - | 3, 4 | 400 | |||
DB498812.1 | - | 3, 4 | 360 | |||
U78170.1 | - | 3, 4 | 333 | |||
Y12336.1 | - | 3, 4 | 426 |