Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289123.1 | NP_001276052.1 | 5 | 1667 | ||
NM_001289127.1 | NP_001276056.1 | 5 | 1649 | |||
NM_001289129.1 | NP_001276058.1 | 5 | 1597 | |||
NM_001289130.1 | NP_001276059.1 | 4 | 1706 | |||
NM_001289131.1 | NP_001276060.1 | 4 | 1673 | |||
NM_006087.3 | NP_006078.2 | 4 | 1749 | |||
GenBank mRNA | AK075307.1 | - | 4 | 1475 | ||
AK293503.1 | - | 4 | 1398 | |||
AK294102.1 | - | 3 | 850 | |||
AK295917.1 | - | 5 | 1228 | |||
BC006570.2 | - | 4 | 1458 | 3603726 | ||
BC013683.1 | - | 4 | 1484 | 4155617 |