Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204453.1 | NP_001191382.1 | 4, 5 | 486 | ||
NM_002803.3 | NP_002794.1 | 4, 5 | 486 | |||
XM_005250505.1 | XP_005250562.1 | 5, 6 | 607 | |||
GenBank mRNA | AB075520.1 | - | 4, 5 | 347 | ||
AK298529.1 | - | 4, 5 | 334 | |||
AK312648.1 | - | 4, 5 | 338 | |||
BC002589.2 | - | 4, 5 | 330 | 3161790 | ||
D11094.1 | - | 4, 5 | 364 |