Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006398.3 | NP_006389.2 | 2 | 307 | ||
GenBank mRNA | AF123050.1 | - | 2 | 307 | ||
AK311914.1 | - | 2 | 114 | |||
BC012472.1 | - | 2 | 107 | 4517233 | ||
DB181897.1 | - | 2 | 218 | |||
Y12653.1 | - | 2 | 101 |