Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006503.3 | NP_006494.1 | 3 | 446 | ||
NM_153001.2 | NP_694546.1 | 3 | 353 | |||
GenBank mRNA | AF020736.1 | - | 3 | 288 | ||
AF038965.1 | - | 3 | 270 | |||
AK290285.1 | - | 3 | 254 | |||
AK310135.1 | - | 3 | 271 | |||
AK313183.1 | - | 3 | 288 | |||
BC000343.2 | - | 3 | 268 | 2823007 | ||
BC010396.1 | - | 3 | 192 | 4046205 | ||
BC014488.1 | - | 3 | 268 | 4899042 | ||
BC018811.2 | - | 3 | 268 | |||
BI828251.1 | - | 3 | 446 | |||
BT007232.1 | - | 3 | 248 | |||
U27515.1 | - | 3 | 260 |