Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006822.2 | NP_006813.1 | 6 | 1449 | ||
XM_005256334.4 | XP_005256391.1 | 6 | 1652 | |||
XM_006722271.3 | XP_006722334.1 | 6 | 1218 | |||
XM_011523528.2 | XP_011521830.1 | 6 | 1571 | |||
XM_017024042.1 | XP_016879531.1 | 7 | 2084 | |||
XM_017024043.1 | XP_016879532.1 | 6 | 1561 | |||
GenBank mRNA | AK095782.1 | - | 1 | 2213 | ||
AK124376.1 | - | 2 | 2065 | |||
BC018039.1 | - | 6 | 1449 | 4801271 | ||
U05227.1 | - | 6 | 1369 |