Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006909.2 | NP_008840.1 | 24, 25 | 3483 | ||
XM_017009682.1 | XP_016865171.1 | 24, 25 | 3165 | |||
GenBank mRNA | AF023130.1 | - | 24, 25 | 3433 | ||
AF181250.1 | - | 24, 25 | 3433 | |||
BC126112.1 | - | 24, 25 | 3483 | 8991828 | ||
BC136296.1 | - | 24, 25 | 3483 | 9020283 |