Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286059.1 | NP_001272988.1 | 5 | 841 | ||
NM_001286060.1 | NP_001272989.1 | 5 | 676 | |||
NM_001286061.1 | NP_001272990.1 | 5 | 574 | |||
NM_014488.4 | NP_055303.2 | 6 | 777 | |||
GenBank mRNA | AF498956.1 | - | 4 | 435 | ||
AK291376.1 | - | 5 | 836 | |||
AK312323.1 | - | 5 | 582 | |||
BC014213.2 | - | 6 | 696 | 4763140 | ||
BP336371.1 | - | 5 | 573 | |||
BX641138.1 | - | 5 | 621 | |||
CR542106.1 | - | 4 | 435 | |||
U57092.1 | - | 4 | 453 |