Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014709.3 | NP_055524.3 | 3 | 558 | ||
GenBank mRNA | AB011142.2 | - | 4 | 613 | ||
AJ586138.1 | - | 1 | 83 | |||
BC141839.1 | - | 4 | 613 | |||
BX473680.1 | - | 3 | 564 |