Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287059.1 | NP_001273988.1 | 3 | 632 | ||
NM_015052.4 | NP_055867.3 | 4 | 867 | |||
XM_005249665.3 | XP_005249722.1 | 3 | 646 | |||
XM_006715670.3 | XP_006715733.1 | 2 | 544 | |||
XM_006715671.3 | XP_006715734.1 | 2 | 544 | |||
XM_006715673.3 | XP_006715736.1 | 2 | 544 | |||
XM_011515220.2 | XP_011513522.1 | 3 | 437 | |||
XM_011515222.2 | XP_011513524.1 | 2 | 578 | |||
XM_011515223.2 | XP_011513525.1 | 2 | 379 | |||
XM_011515224.2 | XP_011513526.1 | 2 | 859 | |||
XM_017011882.1 | XP_016867371.1 | 2 | 886 | |||
XM_017011883.1 | XP_016867372.1 | 3 | 823 | |||
XM_017011884.1 | XP_016867373.1 | 2 | 886 | |||
XM_017011885.1 | XP_016867374.1 | 4 | 819 | |||
XM_017011886.1 | XP_016867375.1 | 3 | 651 | |||
XM_017011887.1 | XP_016867376.1 | 2 | 886 | |||
XM_017011888.1 | XP_016867377.1 | 2 | 886 | |||
XM_017011889.1 | XP_016867378.1 | 3 | 943 | |||
XM_017011890.1 | XP_016867379.1 | 3 | 651 | |||
GenBank mRNA | AB002320.2 | - | 4 | 867 | ||
AB048365.1 | - | 1 | 219 | |||
AK090799.1 | - | 2 | 886 | |||
AK294918.1 | - | 3 | 586 | |||
BC151227.1 | - | 4 | 867 | 9007236 |