Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016131.4 | NP_057215.3 | 6 | 2163 | ||
GenBank mRNA | AF106681.1 | - | 6 | 1509 | ||
AK023223.1 | - | 6 | 1913 | |||
AK025136.1 | - | 1 | 694 | |||
AL136650.1 | - | 6 | 1922 |